Learn More
Torrey Pines Scientific™ EchoTherm™ Benchtop Chilling/Heating Incubator Sample Blocks
Catalog No. 1168027
Change view
Click to view available options
Holds:
0.2mL tubes
0.5mL/1.5mL tubes
1.5mL tubes
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
11-680-27 0.2mL tubes
11-680-26 0.5mL/1.5mL tubes
11-680-25 1.5mL tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-680-27 Supplier Torrey Pines Scientific™ Supplier No. 6205015
Add to Cart
Add to Cart
Blocks are machined from solid aluminum to fit precisely the tube size and shape. Block bottoms are also machined to nest on the incubator plate to prevent movement. Feature an anodized black finish for better heat transfer.
Specifications
For Use With (Equipment) Echotherm benchtop chilling/heating incubator
Holds 0.2mL tubes
Material Aluminum
Type Block
For Use With (Application) For 0.2mL centrifuge tubes spaced for 8 and 12 tube strip