Learn More
Torrey Pines Scientific™ EchoTherm™ Benchtop Chilling/Heating Incubator Sample Blocks
Catalog No. 1168031
Change view
Click to view available options
Material:
Aluminum
Catalog No. Material
11-680-31 Aluminum
1 options
Catalog No. 11-680-31 Supplier Torrey Pines Scientific™ Supplier No. 6205020
Add to Cart
Add to Cart
Blocks are machined from solid aluminum to fit precisely the tube size and shape. Block bottoms are also machined to nest on the incubator plate to prevent movement. Feature an anodized black finish for better heat transfer.
Specifications
For Use With (Equipment) Echotherm benchtop chilling/heating incubator
Material Aluminum
Type Block
For Use With (Application) Flat bottom 96 and 384-well Assay Plates