Learn More
Research Products International Corp Echotherm Benchtop Incubator
SDP
Catalog No. 501976816
Encompass
Ideal benchtop size incubators provide exact temperatures control to 0.1°C. Store up to three individual programs for instant recall as needed. Each program can include up to ten steps. Modern peltier electronics allows energy efficient operation. Aluminum housing includes a combination two door system that allows viewing of contents without altering chamber temperature.
Additional features includes:
Electronic calibration
Digital readout
Timer with alarm
Power interrupt protection
Auto-shutoff
Twelve month warranty
Catalog No. 50-197-6816 Supplier Research Products International Corp Supplier No. 254635
Add to Cart
Add to Cart
Request a Quote
Ideal benchtop size incubators provide exact temperatures control to 0.1°C. Store up to three individual programs for instant recall as needed. Each program can include up to ten steps. Modern peltier electronics allows energy efficient operation. Aluminum housing includes a combination two door system that allows viewing of contents without altering chamber temperature.
Additional features includes:
Electronic calibration
Digital readout
Timer with alarm
Power interrupt protection
Auto-shutoff
Twelve month warranty