Learn More
Research Products International Corp EchoTherm Benchtop Chilling/Heating Incubator with Non-Programmable Control
SDP
Catalog No. 501976812
Encompass
The EchoTherm bench-top, solid-state convection chilling/heating incubators are reliable, accurate and easy to use. Peltier-based for both heating and chilling, the units have no compressors or CFCs, making them truly energy efficient and vibration free. The incubator has a 55-liter volume. This model is not programmable.
Catalog No. 50-197-6812 Supplier Research Products International Corp Supplier No. 248707
Add to Cart
Add to Cart
Request a Quote
The EchoTherm bench-top, solid-state convection chilling/heating incubators are reliable, accurate and easy to use. Peltier-based for both heating and chilling, the units have no compressors or CFCs, making them truly energy efficient and vibration free. The incubator has a 55-liter volume. This model is not programmable.