Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Torrey Pines Scientific™ EchoTherm™ Benchtop Incubator, Chiller and Freezer

Catalog No. 1168022
Change view
Click to view available options
Temperature Range (Metric):
-10°C to +100°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Temperature Range (Metric)
11-680-22 -10°C to +100°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-680-22 Supplier Torrey Pines Scientific™ Supplier No. IC20
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Includes RS-232-C interface for computer-directed data collection and unit control.A built-in datalogger can be programmed to collect temperature data points in 1-second, 1-minute, or 5-minute intervals to a total of 8110 data points. Includes a universal power supply that will accept inputs from 85 to 265 VAC, 50/60Hz and 12VDC output to chiller/heater module.

Five devices in one—water bath, dry bath, incubator, refrigerator, and freezer; Heats, chills, or freezes samples from -10° to +100°C—even at ambient temperatures; Energy-efficient Peltier design—no compressor or CFCs.

Compact, easy-to-use, and energy-efficient, Echotherm units are ideal for field or benchtop applications. Digital control from –10° to +100°C with ±1°C accuracy. Electronic calibration is factory set, but can be easily recalibrated to suit the user's needs. Two-line, alphanumeric, digital display is backlit for easy reading, shows target and actual temperatures continuously, and timer settings on demand. Integral countdown timer has a range from 1 second to 30 days, an audible alarm, and a user-settable auto-off feature. A buffer lamp on the front panel of the unit flashes when the datalogger buffer is collecting data and stays lit when the buffer is full of data to be downloaded. 50-watt power consumption.

  • RS-232-C interface for computer-directed data collection and unit control
  • Built-in datalogger collects temperature data points in 1-sec., 1-minute, or 5-minute intervals to a total of 8110 data points

Order Info

Small footprint, 8 3/4L x 6 1/2W x 3 1/2 in.H (22.2 x 16.5 x 8.9cm), takes up less bench space and is easily transported for remote applications. Aluminum sample blocks and clear polyethylene sample covers are available separately. Net weight (unit and power supply): 6.5 lb. (2.9kg). Aluminum chilling/heating surface measures 2 7/8 x 4 3/8 in. (7.3 x 11.1cm) and is very flat to facilitate uniform temperature transfer to samples.

Certifications

UL/CSA/CE approved.

Specifications

Certifications/Compliance UL, CSA, CE
Material Aluminum
Temperature Range (Metric) -10°C to +100°C
Type Dry Bath
Display Digital
Includes RS232C interface, built-in datalogger, universal power supply, digital control, Integral countdown timer, Buffer lamp, audible alarm
Model IC20
Run Time 00:01 sec. to 30 days
Temperature Accuracy ±1°C
Temperature Uniformity ±0.3°C
Warranty 1 Year
Dimensions (L x W x H) 6.5 x 2.2 x 3.2 in. (16.5 x 5.6 x 8.1 cm)
Length (English) 6.5 in.
Length (Metric) 16.5 cm
Width (English) 2.2 in.
Width (Metric) 5.6 cm
Height (English) 3.2 in.
Height (Metric) 8.1 cm
Weight (English) 6.5 lb.
Weight (Metric) 2.9 kg
Electrical Requirements 12 V, 50/60 Hz
Voltage 12 V
Wattage 50 w
Frequency 50/60 Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.