Learn More
Research Products International Corp Echotherm Programmable Benchtop Incubator, 27.5 Liter Volume, Timer Range 99 Hours
SDP
Catalog No. 50212599
Encompass
This benchtop chilling incubator is the ideal laboratory instrument for performing enzyme reactions, deactivations and many other procedures that require heating or chilling of samples. Peltier based heating and cooling functions are energy efficient. Solid state controls provide high reliability and accuracy and feature digital display and setting. Built-in safety features include self diagnostics and special circuitry to eliminate control errors.
Echotherm is constructed with a metal chassis and an ABS plastic front panel and door. A separate clear plastic inner door allows viewing without disrupting chamber temperature. Seamless chamber walls are easy to clean.
Supplied with two racks, 3-wire grounded line cord and instruction manual.
Catalog No. 50-212-599 Supplier Research Products International Corp Supplier No. 248720
This benchtop chilling incubator is the ideal laboratory instrument for performing enzyme reactions, deactivations and many other procedures that require heating or chilling of samples. Peltier based heating and cooling functions are energy efficient. Solid state controls provide high reliability and accuracy and feature digital display and setting. Built-in safety features include self diagnostics and special circuitry to eliminate control errors.
Echotherm is constructed with a metal chassis and an ABS plastic front panel and door. A separate clear plastic inner door allows viewing without disrupting chamber temperature. Seamless chamber walls are easy to clean.
Supplied with two racks, 3-wire grounded line cord and instruction manual.