Learn More
Research Products International Corp Echotherm Digital Hot Plate and Stirrer
SDP
Catalog No. 501976889
Encompass
EchoTherm Digital Hot Plate/Stirrer features constant display of all functions. LCD display functions include display target and actual plate temperature: target and actual solution temperature stirring speed; temperature ramping; and count down timer with auto/off.
Easy to use membrane keyboard allows heating, stirring, timing and electronic calibration. Chemically resistant ceramic glass top heats quickly. Ceramic top is mounted on aluminum frame and provides cool edge to prevent burns. Platinum RTD and PID temperature measurement systems enable precise and accurate temperature control.
Supplied with 6-ft grounded line cord, fuse holder, and main on/off switch.
Catalog No. 50-197-6889 Supplier Research Products International Corp Supplier No. 249853
EchoTherm Digital Hot Plate/Stirrer features constant display of all functions. LCD display functions include display target and actual plate temperature: target and actual solution temperature stirring speed; temperature ramping; and count down timer with auto/off.
Easy to use membrane keyboard allows heating, stirring, timing and electronic calibration. Chemically resistant ceramic glass top heats quickly. Ceramic top is mounted on aluminum frame and provides cool edge to prevent burns. Platinum RTD and PID temperature measurement systems enable precise and accurate temperature control.
Supplied with 6-ft grounded line cord, fuse holder, and main on/off switch.