Learn More
Research Products International Corp EchoTherm Dual Station Chilling/Heating Plate
SDP
Catalog No. 501976845
Encompass
Dual Station Chilling/Heating Plate allows two blocks to be run at different temperatures. Two separate timers and data loggers can be individually controlled to provide maximum versatility and efficiency. Collects up to 8110 data points per station at 1 second, 1 minute or 5 minute intervals and includes a built in RS232 I/O port. Audible alarm signals for one minute upon completion or run. Timer counts up until unit is turned off. Unit can be set to automatically turn off at a pre-selected time.
Ideal for a variety of molecular biology applications such as storing enzymes or DNA libraries at a workstation; ligation reactions; maintaining samples at ice bath temperature and more. Surface mounted platforms accommodate 96-well assay plates or standard aluminum blocks (2 7/8 x 4 3/8 Inches).
Supplied with AC line cord, instruction manual and twelve month warranty.
Catalog No. 50-197-6845 Supplier Research Products International Corp Supplier No. 248885
Dual Station Chilling/Heating Plate allows two blocks to be run at different temperatures. Two separate timers and data loggers can be individually controlled to provide maximum versatility and efficiency. Collects up to 8110 data points per station at 1 second, 1 minute or 5 minute intervals and includes a built in RS232 I/O port. Audible alarm signals for one minute upon completion or run. Timer counts up until unit is turned off. Unit can be set to automatically turn off at a pre-selected time.
Ideal for a variety of molecular biology applications such as storing enzymes or DNA libraries at a workstation; ligation reactions; maintaining samples at ice bath temperature and more. Surface mounted platforms accommodate 96-well assay plates or standard aluminum blocks (2 7/8 x 4 3/8 Inches).
Supplied with AC line cord, instruction manual and twelve month warranty.