Learn More
Research Products International Corp EchoTherm Electronic Chilling/Heating Plate, Digital
SDP
Catalog No. 501976894
Encompass
Ideal bench top chiller/freezer/incubator for biological samples. Cools or heats samples from -10°C to 100°C with a controlled accuracy of +/-1°C. Features a built-in data logger with 8110 data points collectable in 1 second, 1 minute or 5 minute intervals and RS232 port.
Accommodates 96-well assay plates and most brands of aluminum blocks that hold 0.2 ml, 0.5 ml, or 1.5 ml-2.0 ml micro-tubes. Surface mounted chilling plate allows convenient access by robotic systems or for use as a chilling/heating microscope stage.
Compact size (6 1/2 x 8 3/4 x 3 1/2 Inches) takes up a minimal amount of bench space. Operates on 12 volts DC from an external power supply that is compliant with UL, CSA and CE specifications.
Supplied with universal power supply, interconnect cable, instruction manual and 12 month warranty.
Catalog No. 50-197-6894 Supplier Research Products International Corp Supplier No. 248807
Ideal bench top chiller/freezer/incubator for biological samples. Cools or heats samples from -10°C to 100°C with a controlled accuracy of +/-1°C. Features a built-in data logger with 8110 data points collectable in 1 second, 1 minute or 5 minute intervals and RS232 port.
Accommodates 96-well assay plates and most brands of aluminum blocks that hold 0.2 ml, 0.5 ml, or 1.5 ml-2.0 ml micro-tubes. Surface mounted chilling plate allows convenient access by robotic systems or for use as a chilling/heating microscope stage.
Compact size (6 1/2 x 8 3/4 x 3 1/2 Inches) takes up a minimal amount of bench space. Operates on 12 volts DC from an external power supply that is compliant with UL, CSA and CE specifications.
Supplied with universal power supply, interconnect cable, instruction manual and 12 month warranty.