Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Torrey Pines Scientific™ EchoTherm™ IN45 Benchtop Refrigerated Incubator, 55 L, ABS Plastic

Catalog No. 1168037
Change view
Click to view available options
Capacity (Metric):
55 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
11-680-37 55 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. 11-680-37 Supplier Torrey Pines Scientific™ Supplier No. IN45
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

Temperature Range: 4°C to 70°C

The IN30 and IN40 Series bench-top, solid-state convection chilling/heating incubators are reliable, accurate and easy to use. Peltier-based for both heating and chilling, the units have no compressors or CFCs, making them truly energy efficient. The IN30 Series has 27.5-liter volume and the IN40 Series has 55L volume. Neither the IN30 and IN40 are programmable, however, the IN35 and IN45 are fully programmable, but can also be used in a non-programmable mode.

  • Peltier-based, no compressors, no CFCs, energy-efficient
  • Temperature range from 4° to 70°C
  • Fully programmable or non-programmable controls
  • 27.5 or 55L volumes
  • Timer with alarm and Auto-Off
  • Electronic calibration and power failure protection
  • RS-232 interface
Specifications
  • Range: 4.0° to 70.0°C (IN40/IN45 max cold is 16°C below room temperature)
  • Readability: ±0.1°C
  • Accuracy: ±0.2°C
  • Stability: ±0.1°C
  • Sensor Type: Solid State IC
  • Controller Type: PID
  • Chamber Uniformity: ±0.5°C
  • Convection Fan: Yes

  • Protein crystal growth
  • BODs
  • Culture growth above, below or at ambient
  • Enzyme reactions and deactivations
  • Incubating marine cultures below ambient
  • Ligations at 14° to 16°C
  • Hybridizations
  • Storing oocytes at 17°C
  • Storing DNA libraries

Compliance

UL, CSA , and CE listed

Warranty and Services

One year

Specifications

Capacity (English) 1.94 cu. ft.
Certifications/Compliance UL, CSA, CE
Capacity (Metric) 55 L
Chamber Material ABS Plastic
Temperature Range (Metric) 4°C to 70°C
Type Refrigerated Incubator
Display LCD
Model IN45
Temperature Accuracy ±0.1°C
Warranty 1 Year
Width (English) 22.25 in.
Width (English) Interior 14 in.
Width (Metric) 56.5 cm
Width (Metric) Interior 35.6 cm
Height (English) 28.25 in.
Height (English) Interior 24 in.
Height (Metric) 71.75 cm
Height (Metric) Interior 61 cm
Diameter (English) Inside 10 in.
Diameter (English) Outside 18.5 in.
Diameter (Metric) Inside 64.5 cm
Diameter (Metric) Outside 47 cm
Weight (English) 56 lb.
Weight (Metric) 25.3 kg
Electrical Requirements 12 V, 50/60 Hz
Voltage 12 V
Frequency 50/60 Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.