Learn More
Research Products International Corp EchoTherm Orbital Chilling/Heating Dry Bath, Non-Programmable
SDP
Catalog No. 501976833
Encompass
The ideal benchtop tool for all Molecular Biology applications. This compact digital dry bath provides quick and accurate chilling or heating from -10°C to 100° and orbital mixing from 200 to 1000 rpm. Solid state electronics and Peltier design provides the highest reliability and energy efficient performance. Offered in both programmable and non-programmable models. The programmable unit stores up to five programs with 10 steps per program. Programs can be automatically repeated 1 to 99 times. Built in data logger stores 7825 data points collectable in 1 second, or 1 and 5 minute intervals. The non-programmable model also includes a built-in data logger with 8110 data point capability.
Catalog No. 50-197-6833 Supplier Research Products International Corp Supplier No. 348252
The ideal benchtop tool for all Molecular Biology applications. This compact digital dry bath provides quick and accurate chilling or heating from -10°C to 100° and orbital mixing from 200 to 1000 rpm. Solid state electronics and Peltier design provides the highest reliability and energy efficient performance. Offered in both programmable and non-programmable models. The programmable unit stores up to five programs with 10 steps per program. Programs can be automatically repeated 1 to 99 times. Built in data logger stores 7825 data points collectable in 1 second, or 1 and 5 minute intervals. The non-programmable model also includes a built-in data logger with 8110 data point capability.