Learn More
Applied Biophysics Inc ECIS CULTURE ARRAYS 8WELL 6/PK
SDP
Catalog No. NC0625802
Encompass
  • Disposable electrode arrays for real-time monitoring of cellular behaviors
  • Measures impedance changes across adherent cell layers
  • Designed for Electric Cell-substrate Impedance Sensing (ECIS) applications
  • Uses transparent polycarbonate substrate for microscopic observation
  • 8 wells per array with 40 circular active electrodes
  • Electrode configuration features two sets of inter-digitated fingers per well
  • Active electrode diameter is 250 µm
  • Electrode area per well is 1.96 mm²
  • Maximum well volume is 600 µL
  • Approximately 2000 to 4000 cells can be measured per well
  • Substrate thickness is 0.51 mm
  • Packaged 6 per pack
Catalog No. NC0625802 Supplier Applied Biophysics Inc Supplier No. 8W10E+ PC
  • Disposable electrode arrays for real-time monitoring of cellular behaviors
  • Measures impedance changes across adherent cell layers
  • Designed for Electric Cell-substrate Impedance Sensing (ECIS) applications
  • Uses transparent polycarbonate substrate for microscopic observation
  • 8 wells per array with 40 circular active electrodes
  • Electrode configuration features two sets of inter-digitated fingers per well
  • Active electrode diameter is 250 µm
  • Electrode area per well is 1.96 mm²
  • Maximum well volume is 600 µL
  • Approximately 2000 to 4000 cells can be measured per well
  • Substrate thickness is 0.51 mm
  • Packaged 6 per pack