Learn More
Abcam ECL Substrate Kit (High Sensitivity), 200ML
Catalog No. NC1791705
Encompass_Preferred
High Sensitivity ECL Substrate Kit ab133406 is designed for the detection of proteins with 23pg-187ng of protein per band.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1791705 Supplier Abcam Supplier No. AB133406200ML
High Sensitivity ECL Substrate Kit ab133406 is designed for the detection of proteins with 23pg-187ng of protein per band.
The product is subject to the following: Abcam Restricted Use Statement