Learn More
Abcam ECL Substrate Kit (Ultra High Sensitivity), 100ML
Catalog No. 502583981
Encompass_Preferred
Ultra High Sensitivity ECL Substrate Kit ab133409 is a horseradish peroxidase (HRP) substrate specially developed for Western blotting detection of very low abundance proteins with 1.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3981 Supplier Abcam Supplier No. AB133409100ML
Ultra High Sensitivity ECL Substrate Kit ab133409 is a horseradish peroxidase (HRP) substrate specially developed for Western blotting detection of very low abundance proteins with 1.
The product is subject to the following: Abcam Restricted Use Statement