Learn More
Abcam ECL Western Blotting Substrate Kit, 50 Tests
Catalog No. 502204750
Encompass_Preferred
The ECL Western Blotting Substrate is a highly sensitive, nonradioactive, enhanced luminol-based chemiluminescent substrate for easy detection of horseradish peroxidase (HRP) on immunoblots.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-4750 Supplier Abcam Supplier No. AB6562350TEST
Add to Cart
Add to Cart
The ECL Western Blotting Substrate is a highly sensitive, nonradioactive, enhanced luminol-based chemiluminescent substrate for easy detection of horseradish peroxidase (HRP) on immunoblots.
The product is subject to the following: Abcam Restricted Use Statement