Learn More
Labcon North America Eclipse™ 1200uL Pipet Tips for Rainin LTS Pipettors, in Eclipse™ Refill
Catalog No. NC1166735
Encompass_Preferred
Tip, Pipet; Labcon; Eclipse™; Specialty; 1200uL; Fits 1200uL Rainin LTS style pipettors; Low mounting and ejection forces less than 3kg; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; 960 tips per low waste Eclipse™ Refill; 10 refills per case; 88.9mm. H; Made in USA
Catalog No. NC1166735 Supplier Labcon North America Supplier No. 10192600009
Tip, Pipet; Labcon; Eclipse™; Specialty; 1200uL; Fits 1200uL Rainin LTS style pipettors; Low mounting and ejection forces less than 3kg; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; 960 tips per low waste Eclipse™ Refill; 10 refills per case; 88.9mm. H; Made in USA