Learn More
Labcon North America Labcon Eclipse 200 µL Pipet Tips, 100% Virgin Polypropylene, Certified RNase/DNase Free, 9600 Tips/Case
Catalog No. NC2100791
Encompass_Preferred
Labcon Eclipse 200 µL Graduated Pipet Tips provide precision and accuracy in liquid handling. Made from 100% pure virgin medical-grade polypropylene, they are certified free of RNase/DNase, ATP, and endotoxin contamination. Designed with a graduated format for easy volume verification and beveled points to reduce fluid retention, these autoclavable tips fit most popular 200 µL pipettors, making them an excellent choice for reliable lab results and efficient waste management.
This item is not returnable. View return policy
Catalog No. NC2100791 Supplier Labcon North America Supplier No. 10932600009
Labcon Eclipse 200 µL Graduated Pipet Tips provide precision and accuracy in liquid handling. Made from 100% pure virgin medical-grade polypropylene, they are certified free of RNase/DNase, ATP, and endotoxin contamination. Designed with a graduated format for easy volume verification and beveled points to reduce fluid retention, these autoclavable tips fit most popular 200 µL pipettors, making them an excellent choice for reliable lab results and efficient waste management.