Learn More
MSC Eclipse Magnetics Lifting Magnet, 550 lb Flat SWL, Neodymium, Safety Shim
Catalog No. 5000529462
Encompass
The Eclipse Magnetics Ultralift Plus Lifting Magnet UL0550+ is a powerful, permanently magnetic device designed for efficient and safe lifting of various ferrous loads. It uses high-intensity neodymium rare earth magnets and features manual on/off switching, requiring no external power. Multiple built-in safety mechanisms ensure secure operation.
  • Lifting eye sensor prevents accidental release when under tension.
  • Patented safety shim allows pre-testing to ensure a 3:1 safety factor.
  • Safety button locks handle in "ON" position, preventing accidental load release.
  • Does not mark the load surface.
  • Space-saving design requires access only to the top of the load.
  • Easy to install and suitable for one-person operation.
Catalog No. 50-005-29462 Supplier MSC Supplier No. 04891685
The Eclipse Magnetics Ultralift Plus Lifting Magnet UL0550+ is a powerful, permanently magnetic device designed for efficient and safe lifting of various ferrous loads. It uses high-intensity neodymium rare earth magnets and features manual on/off switching, requiring no external power. Multiple built-in safety mechanisms ensure secure operation.
  • Lifting eye sensor prevents accidental release when under tension.
  • Patented safety shim allows pre-testing to ensure a 3:1 safety factor.
  • Safety button locks handle in "ON" position, preventing accidental load release.
  • Does not mark the load surface.
  • Space-saving design requires access only to the top of the load.
  • Easy to install and suitable for one-person operation.