Learn More
Electron Microscopy Sciences NIGHTSEA™ Eclipse MicroTent
SDP
Catalog No. NC1507737
Encompass_Preferred
The Eclipse MicroTent provides local darkness around conventional laboratory stereo microscopes that are using a fluorescence adapter. Fluorescence can be weak and can be difficult to see well if there is ambient light. The tent is ideal for sheilding microscopes on lab benches in shared spaces, near windows, or in other difficult-to-darken locations such as in the field. Easy access to the sample stage and the focus and zoom controls is maintained. It is designed for stereo microscopes but could potentially be used with many varieties of compound microscopes.
Catalog No. NC1507737 Supplier Electron Microscopy Sciences Supplier No. SFATENT
The Eclipse MicroTent provides local darkness around conventional laboratory stereo microscopes that are using a fluorescence adapter. Fluorescence can be weak and can be difficult to see well if there is ambient light. The tent is ideal for sheilding microscopes on lab benches in shared spaces, near windows, or in other difficult-to-darken locations such as in the field. Easy access to the sample stage and the focus and zoom controls is maintained. It is designed for stereo microscopes but could potentially be used with many varieties of compound microscopes.