Learn More
Med Vet International Becton Dickinson Syringe & Needle, 3mL, Luer Lock, 25G X 5/8", Hypodermic, 50/BX, 305781
Catalog No. 503043646
Encompass_Preferred
BD 305781 3mL Syringe & Needle Combo comes with a clear barrel and easy to read graduation marks. The Luer Lock tip of this 3mL Syringe & Needle Combo fits conventional and safety needles. This BD 305781 3mL Syringe & Needle Combo comes with a 25G X 5/8" Thin Wall needle. Thin wall needles are thinner than regular wall needles. The outside diameter is the same size. However, the inside diameter is bigger than the regular wall's inside diameter. Thin wall needles provide more comfort with an increased flow rate. This contains a hypodermic needle which is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 50/BX
Catalog No. 50-304-3646 Supplier Med Vet International Supplier No. 305781
BD 305781 3mL Syringe & Needle Combo comes with a clear barrel and easy to read graduation marks. The Luer Lock tip of this 3mL Syringe & Needle Combo fits conventional and safety needles. This BD 305781 3mL Syringe & Needle Combo comes with a 25G X 5/8" Thin Wall needle. Thin wall needles are thinner than regular wall needles. The outside diameter is the same size. However, the inside diameter is bigger than the regular wall's inside diameter. Thin wall needles provide more comfort with an increased flow rate. This contains a hypodermic needle which is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 50/BX