Learn More
TARGETMOL CHEMICALS INC Ecliptasaponin A 20MG
SDP
Catalog No. 502878928
Encompass_Preferred
Also available in 1 mL and bulk. Please contact Fisher for quotes. 1. Ecliptasaponin A (Echinocystic acid-3-o-glucoside) has protective effects against the pulmonary fibrosis induced by bleomycin via reducing the oxidative stress, lung tissue inflammation, and the subsequent epithelial-mesenchymal transition. Purity 99.71%
Catalog No. 50-287-8928 Supplier TARGETMOL CHEMICALS INC Supplier No. T2S069020MG
Also available in 1 mL and bulk. Please contact Fisher for quotes. 1. Ecliptasaponin A (Echinocystic acid-3-o-glucoside) has protective effects against the pulmonary fibrosis induced by bleomycin via reducing the oxidative stress, lung tissue inflammation, and the subsequent epithelial-mesenchymal transition. Purity 99.71%