Learn More
Medchemexpress LLC Ecliptasaponin D | 206756-04-9 | 5 MG
SDP
Catalog No. 5000418223
Encompass_Preferred
Ecliptasaponin D is a triterpenoid glucoside isolated from Eclipta alba (L.) Hassk, which is the aerial part of Eclipta prostrate. Eclipta prostrate is considered a nourishing herbal medicine with pleiotropic effects, including anti-inflammatory, hepatoprotective, antioxidant, and immunomodulatory properties.
  • Triterpenoid glucoside
  • Isolated from Eclipta alba (L.) Hassk
  • Anti-inflammatory properties
  • Hepatoprotective properties
  • Antioxidant properties
  • Immunomodulatory properties
Catalog No. 50-004-18223 Supplier Medchemexpress LLC Supplier No. HYN21915MG
Ecliptasaponin D is a triterpenoid glucoside isolated from Eclipta alba (L.) Hassk, which is the aerial part of Eclipta prostrate. Eclipta prostrate is considered a nourishing herbal medicine with pleiotropic effects, including anti-inflammatory, hepatoprotective, antioxidant, and immunomodulatory properties.
  • Triterpenoid glucoside
  • Isolated from Eclipta alba (L.) Hassk
  • Anti-inflammatory properties
  • Hepatoprotective properties
  • Antioxidant properties
  • Immunomodulatory properties