Learn More
Medchemexpress LLC Ecliptasaponin D | 206756-04-9 | MFCD30724971 | 99.3% | 634.84 | C36H58O9 | 10 MG
SDP
Catalog No. 5000295726
Encompass_Preferred
Ecliptasaponin D is a triterpenoid glucoside isolated from Eclipta alba and supplied for research and analytical applications. It is reported to have anti-inflammatory, hepatoprotective, antioxidant, and immunomodulatory activities. Identifiers include CAS 206756-04-9 and molecular formula C36H58O9.
  • High purity suitable for analytical and screening assays.
  • Characterized molecular weight and formula for reproducibility.
  • Supplied in small milligram quantities for compound screening and method development.
  • Stable when stored at -20°C and protected from light; in solvent store at -80°C for long-term storage.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-95726 Supplier Medchemexpress LLC Supplier No. HYN219110MG
Ecliptasaponin D is a triterpenoid glucoside isolated from Eclipta alba and supplied for research and analytical applications. It is reported to have anti-inflammatory, hepatoprotective, antioxidant, and immunomodulatory activities. Identifiers include CAS 206756-04-9 and molecular formula C36H58O9.
  • High purity suitable for analytical and screening assays.
  • Characterized molecular weight and formula for reproducibility.
  • Supplied in small milligram quantities for compound screening and method development.
  • Stable when stored at -20°C and protected from light; in solvent store at -80°C for long-term storage.
  • Intended for research use only; not for human or clinical use.