Learn More
Sigma Aldrich Fine Chemicals Biosciences ECLTM Prime Western Blotti
Catalog No. 502904343
Encompass
Amersham(TM) ECL(TM) Prime is a highly sensitive reagent characterized by stable signal emission allowing for the possibility of repeated exposures and making it easier to process several blots in the same experimental run. The high intensity of the emitted signals means that proteins can be detected using highly diluted primary and secondary antibodies contributing to low background and reducing consumption of reagents.Amersham(TM) ECL(TM) Prime consolidates and builds on the benefits of Amersham(TM) ECL(TM) Plus and Amersham(TM) ECL(TM) Advance to deliver a detection system that is sensitive stable precisely quantitative across a wide dynamic range of protein levels and conservative in its consumption of expensive antibody reagents.
Catalog No. 50-290-4343 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GERPN2232
Amersham(TM) ECL(TM) Prime is a highly sensitive reagent characterized by stable signal emission allowing for the possibility of repeated exposures and making it easier to process several blots in the same experimental run. The high intensity of the emitted signals means that proteins can be detected using highly diluted primary and secondary antibodies contributing to low background and reducing consumption of reagents.Amersham(TM) ECL(TM) Prime consolidates and builds on the benefits of Amersham(TM) ECL(TM) Plus and Amersham(TM) ECL(TM) Advance to deliver a detection system that is sensitive stable precisely quantitative across a wide dynamic range of protein levels and conservative in its consumption of expensive antibody reagents.