Learn More
Sigma Aldrich Fine Chemicals Biosciences ECLTM Select Western Blott
Catalog No. 502904253
Encompass
Amersham(TM) ECL(TM) Select Western blotting detection reagent with highly intense signal output for detection of medium to very low protein levels.Amersham(TM) ECL(TM) Select is a high sensitivity reagent for chemiluminescent Western blotting detection. High signal intensity makes Amersham(TM) ECL(TM) Select suitable for the most demanding Western blotting applications allowing for detection of minute protein quantities. Its long lasting signals allow for multiple exposures and a convenient time window between experiment and analysis. Signals can be detected with CCD camera or X-ray film.Amersham(TM) ECL(TM) Select typically exhibits a dynamic range that is complementary to Amersham(TM) ECL(TM) Prime and similar to Amersham(TM) ECL(TM) Advance.
Catalog No. 50-290-4253 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GERPN2235
Amersham(TM) ECL(TM) Select Western blotting detection reagent with highly intense signal output for detection of medium to very low protein levels.Amersham(TM) ECL(TM) Select is a high sensitivity reagent for chemiluminescent Western blotting detection. High signal intensity makes Amersham(TM) ECL(TM) Select suitable for the most demanding Western blotting applications allowing for detection of minute protein quantities. Its long lasting signals allow for multiple exposures and a convenient time window between experiment and analysis. Signals can be detected with CCD camera or X-ray film.Amersham(TM) ECL(TM) Select typically exhibits a dynamic range that is complementary to Amersham(TM) ECL(TM) Prime and similar to Amersham(TM) ECL(TM) Advance.