Learn More
Sigma Aldrich Fine Chemicals Biosciences ECLTM Start Western Blotti
Catalog No. 502900690
Encompass
Amersham ECL start is a chemiluminescent detection reagent in the Amersham ECL product family that is recommended for confirmatory analysis and detection of high to medium levels of endogenous proteins. Amersham ECL start has a long signal duration which enables multiple exposures of the membrane plus a convenient time window between experiment and analysis.
Catalog No. 50-290-0690 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GERPN3243
Amersham ECL start is a chemiluminescent detection reagent in the Amersham ECL product family that is recommended for confirmatory analysis and detection of high to medium levels of endogenous proteins. Amersham ECL start has a long signal duration which enables multiple exposures of the membrane plus a convenient time window between experiment and analysis.