Learn More
Sigma Aldrich Fine Chemicals Biosciences ECLTM Western Blotting Rea
Catalog No. 502904451
Encompass
Amersham(TM) ECL(TM) Western Blotting Detection Reagents were the worlds first commercial chemiluminescent reagents for Western blotting and are still the most widely used. They are at least 10-fold more sensitive than colorimetric assays and produce excellent and reliable results every time.These reagents use enhanced luminol-based detection which is suitable for all routine western blotting. ECL(TM) Western Blotting Detection Reagents in combination with Hybond(R) ECL(TM) membranes form a complete SOLUTIONon for Western blot applications.Emission duration max. 2 hoursContents include the followingGERPN2109 62.5 mL each of ECL(TM) Detection Reagents 1 and 2 (1000 cm2)GERPN2209 125 mL each (2000 cm2)GERPN2106 250 mL each (4000 cm2)GERPN2134 375 mL each (6000 cm2).
Catalog No. 50-290-4451 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GERPN2106
Amersham(TM) ECL(TM) Western Blotting Detection Reagents were the worlds first commercial chemiluminescent reagents for Western blotting and are still the most widely used. They are at least 10-fold more sensitive than colorimetric assays and produce excellent and reliable results every time.These reagents use enhanced luminol-based detection which is suitable for all routine western blotting. ECL(TM) Western Blotting Detection Reagents in combination with Hybond(R) ECL(TM) membranes form a complete SOLUTIONon for Western blot applications.Emission duration max. 2 hoursContents include the followingGERPN2109 62.5 mL each of ECL(TM) Detection Reagents 1 and 2 (1000 cm2)GERPN2209 125 mL each (2000 cm2)GERPN2106 250 mL each (4000 cm2)GERPN2134 375 mL each (6000 cm2).