Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTX™ ECM™ 2001+ HV Output Cable

Catalog No. BTX452057 Shop All BTX Harvard Products
Change view
Click to view available options
Length (Metric) Cable:
1.5 m
3 m
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Cable
BTX452057 1.5 m
BTX452058 3 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTX452057 Supplier BTX™ Supplier No. 452057
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ECM™ 2001+ HV Output Cable for use with BTX™ ECM™ 2001+.

  • For connection to electroporation generators

Specifications

For Use With (Application) In-Vivo Drug or Gene Delivery, Gene Therapy, Protein Delivery
For Use With (Equipment) ECM™ 2001+
Product Type Cables and Adapters
Description ECM 2001+ HV Output Cable, 5 ft
Includes Cable Only
Length (English) Cable 5 ft.
Length (Metric) Cable 1.5 m
Quantity 1 Ea.
Sterility Non-sterile
Connection Type Output Cable
Pulse Form AC Sine Wave, DC Square Wave
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.