Learn More
BTX™ ECM™ 2001 Safety Stand for Flatpack
Catalog No. BTX470208 Shop All BTX Harvard Products
Change view
Click to view available options
Connection Type:
Banana Cables
Catalog No. Connection Type
BTX470208 Banana Cables
1 options
Catalog No. BTX470208 Supplier BTX™ Supplier No. 470208
Add to Cart
Add to Cart
ECM™ 2001 Safety Stand for Flatpack for use with ECM™ 2001.
  • Cuvette module plugs directly into generator
  • Holds one cuvette
  • Safety swing shield secures cuvette in holder
  • 4 mm Banana Cables, 4 mm F/F Adapters
Specifications
For Use With (Application) Electroporation
For Use With (Equipment) Legacy ECM™ 2001
Product Line BTX Electroporation
Product Type Electroporation accessory
Description Legacy ECM 2001 Safety Stand for Flatpack Chamber
Includes Safety Stand for Flatpack Only
Sterility Non-sterile
Connection Type Banana Cables
Interface Digital User
Pulse Form AC Sine Wave, DC Square Wave