Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 1893MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: ECM1/792. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Stomach, Lung, or breast tumor. Cellular Staining: Cell surface, cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human ECM1 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: ~80kDa.Chromosome: 1q21.2. Gene Symbol: ECM1. Gene ID: 1893. Ensemble: ENSG00000143369. Uniprot: NP_001189787.
Catalog No. 50-188-4658