Learn More
Sino Biological ECM1 Antibody, Rabbit PAb 100uL
Supplier:  Sino Biological 80574RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat ECM1 (rR ECM1 ; Catalog#80574-R08H; NP_446334.1; Met1-Glu562). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-224-7997