Learn More
Description
- Endothelial cells line the interior surface of blood and lymphatic vessels, forming an interface between circulating blood or lymph in the lumen and the rest of tissue/organ
- Laminin-411 binds to the integrin α6β1, located on the cell surface of pluripotent stem cells and induces endothelial cell differentiation
Specifications
Specifications
| Cell Type | Pluripotent stem cells into endothelial cells |
| Content And Storage | 2° to 8°C, Avoid multiple freeze-thaw cycles and protect from light |
| Form | Liquid |
| Substrate | Laminin substrate |
| Includes | 6 x 175 μg ECMatrix™-411 E8 Laminin Substrate (expressed in CHO-S cells) |
| Packaging | 6 x 175 μg |
| Storage Requirements | 2° to 8°C, Avoid multiple freeze-thaw cycles and protect them from light |
| Substrate Quantity | 0.5 mg/mL in PBS |
For research purposes only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.