Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ ECMatrix™-511 E8 Laminin Substrate

Catalog No. CC160350UMI Shop All MilliporeSigma Products
Change view
Click to view available options
Packaging:
100 mL
2 x 175 μg
6 x 175 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
CC160350UMI 2 x 175 μg
CC160100MMI 100 mL
CC1601050MI 6 x 175 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC160350UMI Supplier MilliporeSigma™ Supplier No. CC160350UG
Add to Cart
Add to Cart

ECMatrix™-511 E8 Laminin Substrate has a xeno-free laminin-511 coating for feeder free pluripotent stem cell cultures

  • Human pluripotent stem cells (ES and iPS cells) express α6β1 as the major integrin species and can be maintained stably and expanded efficiently in feeder-free conditions on culture vessels coated with the binding partner laminin-511
  • Animal-free, xeno-free format: Consistent from lot-to-lot with no prescreening required
  • No plate precoating required: Save time by adding to media while passaging cells
  • Supports single cell passaging without ROCKi: Great for CRISPR editing or clonal isolation
  • Higher adhesion and growth rates: Get to your experiments faster
  • Easy to handle: No chilling of cell culture consumables required
TRUSTED_SUSTAINABILITY

Specifications

Cell Type Pluripotent stem cell cultures, CHO-S derived
Form Liquid
Substrate Laminin
Includes 2 x 175 μg EC Matrix™-511 E8 Laminin Substrate (expressed in CHO-S cells)
Packaging 2 x 175 μg
Storage Requirements Store 2°C to 8°C, Avoid multiple freeze-thaw cycles and protect from light
Substrate Quantity 0.5 mg/mL in PBS

For research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.