Learn More
Cayman Chemical EcnoglutIde 10mg
SDP
Catalog No. 502978449
Encompass_Preferred
A cAMP-biased peptide GLP-1R agonist; selectively induces cAMP accumulation over β-arrestin recruitment (EC50s = 0.018 and 1,300 nM, respectively) without inducing GLP-1R receptor internalization (EC50 = >10,000 nM) in cell-based assays using human GLP-1R; reduces fasting and post-prandial blood glucose levels in db/db mice at 0.15 mg/kg; reduces body weight and food intake in db/db mice at 0.05 mg/kg
Catalog No. 50-297-8449 Supplier Cayman Chemical Supplier No. 4127210 MG
A cAMP-biased peptide GLP-1R agonist; selectively induces cAMP accumulation over β-arrestin recruitment (EC50s = 0.018 and 1,300 nM, respectively) without inducing GLP-1R receptor internalization (EC50 = >10,000 nM) in cell-based assays using human GLP-1R; reduces fasting and post-prandial blood glucose levels in db/db mice at 0.15 mg/kg; reduces body weight and food intake in db/db mice at 0.05 mg/kg