Learn More
Chemglass Life Sciences Funnel, Eco, 4" Top Diameter, Thread Size 38-430, Use CG-1759-10 Waste Container
SDP
Catalog No. CG175906
Encompass_Preferred
Funnel, Eco, 4" Top Diameter, Thread Size 38-430, Use CG-1759-10 Waste Container. The 4¿ Eco Funnel is made of HDPE and has a 450mL fluid capacity. Features an easy to use lid, gasket and positive latch to close the lid and maintain an airtight seal. A filter is also built into the interior to reduce the loss of magnetic stir bars and other items when emptying the contents of a vessel into the funnel. The funnel is equipped with an 11¿ stem to reduce chemical evaporation of hazardous solvents in the lab.
Catalog No. CG175906 Supplier Chemglass Life Sciences Supplier No. CG175906
Funnel, Eco, 4" Top Diameter, Thread Size 38-430, Use CG-1759-10 Waste Container. The 4¿ Eco Funnel is made of HDPE and has a 450mL fluid capacity. Features an easy to use lid, gasket and positive latch to close the lid and maintain an airtight seal. A filter is also built into the interior to reduce the loss of magnetic stir bars and other items when emptying the contents of a vessel into the funnel. The funnel is equipped with an 11¿ stem to reduce chemical evaporation of hazardous solvents in the lab.