Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gasco Carbon dioxide | 124-38-9 | MFCD00011491 | 58L

Catalog No. NC1634645
Encompass_Preferred

Carbon dioxide | 1000ppm | MW: 44.009 | 124-38-9 | MFCD00011491 | 58L

Catalog No. NC1634645 Supplier Gasco Supplier No. 58ES341000
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Carbon dioxide | 1000ppm | MW: 44.009 | 124-38-9 | MFCD00011491 | 58L

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.