Learn More
Cole-Parmer Economical Hemacytometer with Cover Slips
Supplier:  Cole-Parmer 7900100
Encompass
This hemacytometer features high light transmission and brightly illuminated line. Double rulings are visible through a wide range of illumination. Neubauer rulings are 0.1 mm below cover glass. The H-shaped moat offers two counting areas. Fused rhodium on glass provides distinct lines. Two glass coverslips are included.
Catalog No. NC0819260