Learn More
Alkali Scientific Eco-Racks Freezer Upright Stai
SDP
Supplier:  Alkali Scientific UFE562
Encompass_Preferred
For Standard 2 Boxes The Alkali ScientificUpright Stainless Steel Freezer Eco-Rack designed for standard 2in boxes represents a significant advancement in freezer storage solutions combining durability eco-friendliness and practicality Alkali ScientificEco-Racks are built from high-quality corrosion-resistant stainless steel ensuring long-lasting performance in the demanding conditions of ultra-low temperature freezers Key Features Corrosion-Resistant Stainless Steel High-quality stainless steel guarantees durability and resistance to the harsh conditions of ultra-low temperatures while ensuring the racks resistance to corrosion maintaining its integrity over time Eco-Friendly Design Utilizing less stainless steel in their construction these Alkali ScientificEco-Racks are more environmentally friendly compared to traditional freezer racks This reduction in material use does not compromise capacity or performance offering a more sustainable storage solution Reduced Frost Accumulat
Catalog No. 50-302-9055