Learn More
Alkali Scientific Eco-Racks Freezer Upright Stai
SDP
Catalog No. 503028553
Encompass_Preferred
For Standard 3 Boxes The Alkali ScientificUpright Stainless Steel Freezer Eco-Rack specifically designed for standard 3in boxes presents an innovative solution emphasizing eco-friendliness and efficiency in freezer storage These racks are crafted from high-quality corrosion-resistant stainless steel tailored to meet the rigorous demands of ultra-low temperature storage while ensuring an eco-friendly footprint by using less material This approach not only contributes to environmental sustainability but also results in less frost accumulation due to reduced surface area offering both practical and ecological benefits Key Features Corrosion-Resistant Stainless Steel Utilizing high-quality stainless steel these racks are built to resist corrosion ensuring longevity and durability even in the challenging conditions of ultra-low (-80C) freezers Eco-Friendly Design The use of less stainless steel in the construction of these Alkali ScientificEco-Racks not only makes them more environment
Catalog No. 50-302-8553 Supplier Alkali Scientific Supplier No. UFE423
For Standard 3 Boxes The Alkali ScientificUpright Stainless Steel Freezer Eco-Rack specifically designed for standard 3in boxes presents an innovative solution emphasizing eco-friendliness and efficiency in freezer storage These racks are crafted from high-quality corrosion-resistant stainless steel tailored to meet the rigorous demands of ultra-low temperature storage while ensuring an eco-friendly footprint by using less material This approach not only contributes to environmental sustainability but also results in less frost accumulation due to reduced surface area offering both practical and ecological benefits Key Features Corrosion-Resistant Stainless Steel Utilizing high-quality stainless steel these racks are built to resist corrosion ensuring longevity and durability even in the challenging conditions of ultra-low (-80C) freezers Eco-Friendly Design The use of less stainless steel in the construction of these Alkali ScientificEco-Racks not only makes them more environment