Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZING™ Enterprises Eco Safety Sign, BioHazard No Eating, Smoking, Drinking Sign, Applying
Description
Plastic signs
- Made from a minimum of 70% post-consumer recycled materials
- Printed with UV-resistant inks
- Pre-drilled for easy mounting
- Include adhesive mounting tabs
- Impact-resistant plastic is suitable for indoor or outdoor use.
Aluminum signs
- Made from 94% recycled materials
- Printed with UV-resistant inks
- Suitable for indoor and outdoor use
- Pre-drilled for easy mounting.
Adhesive signs
- Made from a minimum of 70% post-industrial polystyrene
- Resistant to solvents, moisture, scuffing, tears and UV exposure.
All Zing signs are made in the USA from recycled materials. Recycled materials support sustainability initiatives and LEED credits
Certifications
29 CFR 1910.1030(g)(1)
Specifications
Specifications
| Material | Recycled Polystyrene |
| Quantity | 1 Ea. |
| Mounting Type | Self-adhesive |
| For Use With (Application) | Communication |
| Height (English) | 7 in. |
| Width (English) | 10 in. |
| Languages | English |
| Dimensions (L x W) | 7 x 10 in. |
| Displayed Text | BIOHAZARD EATING, DRINKING, SMOKING, APPLYING COSMETICS AND HANDLING CONTACT LENSES ARE PROHIBITED IN THIS AREA |
| Color | Black on Orange |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?