Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZING™ Enterprises Eco Safety Sign, Warning BioHazard

Catalog No. 18002042
Encompass
Encompass_Preferred
Change view
Click to view available options
Dimensions (L x W):
5 x 7 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
18-002-042 5 x 7 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-002-042 Supplier ZING™ Enterprises Supplier No. 1916S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ZING Enterprises Eco Safety Sign can be applied directly to any hard, flat surface. Available in two sizes.

Adhesive Labels are made from a minimum of 70% post-industrial polystyrene, and are resistant to solvents, moisture, scuffing, tears and UV exposure.

Certifications

29 CFR 1910.1030(g)(1)

Specifications

Material Polystyrene
Quantity 2/Pk.
Mounting Type Self-adhesive
For Use With (Application) Communication
Height (English) 5 in.
Width (English) 7 in.
Languages English
Dimensions (L x W) 5 x 7 in.
Displayed Text BIOHAZARD NO FOOD OR DRINK TO BE STORED IN THIS REFRIGERATOR
Product Type Adhesive Label
Color Black on Orange
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.