Learn More
Zing™ Enterprises Glove Box Dispenser Magnet Mount
Catalog No. 18002993
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
18-002-993 1 Ea.
1 options
This item is not returnable. View return policy
Catalog No. 18-002-993 Supplier Zing™ Enterprises Supplier No. 7226
This item is not returnable. View return policy
Magnets easily attach glove box dispensers to metal surfaces such as filing cabinets, storage cabinets, and metal doors.
  • Designed for use with Zing Glove Box Dispensers
  • Allows dispensers to be easily attached to a variety of metal surfaces
  • Each style of dispenser requires a different amount of magnets for secure mounting:
    • Single - 1 Magnet
    • Double - 2 Magnets
    • Triple - 3 Magnets
Specifications
Color Black
Holds Disposable Glove Boxes
Mounting Magnetic Mount
Quantity 1 Ea.
Thickness (English) 0.125 in.