Learn More
Andwin Scientific ECO Spatula, Double-Ended, with Spoon & Scoop, 14 cm, 300/pk
SDP
Catalog No. NC1540598
Encompass_Preferred
Economical, disposable tools for transferring, collecting, and handling chemical and biological materials. Versatile double-ended design enables use as a spatula, spoon, scoop, scraper, stirring rod, or policeman. The V design at the neck of the disposable laboratory spatula is a trademark of LevGo, Inc
Catalog No. NC1540598 Supplier Andwin Scientific Supplier No. 89233128
Economical, disposable tools for transferring, collecting, and handling chemical and biological materials. Versatile double-ended design enables use as a spatula, spoon, scoop, scraper, stirring rod, or policeman. The V design at the neck of the disposable laboratory spatula is a trademark of LevGo, Inc