Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ECODYST INC ECOCHYLL X1 HYDROGEN WORKHORSE

Supplier:  ECODYST INC ECO777120

Encompass

  • Proprietary direct self-cooling condenser technology
  • No dry ice, liquid nitrogen, water, or glycol needed
  • Minimum condenser temperature of -40 °C
  • Cool-down time to -40 °C in 60 seconds
  • Cool-down time to -20 °C in 90 seconds
  • Faster rates of evaporation
  • Small footprint, under 1 sq. ft.
  • Energy efficiency approximately one-quarter the electricity of a comparable recirculating chiller
  • Compatible with any brand of benchtop rotary evaporator
  • Plug-and-play operation with minimal setup and maintenance
  • Heating bath temperature range from room temperature to 180 °C
  • Rotation speed range from 20 to 280 rpm
  • Large condenser surface area for cooling
  • Eco-friendly and sustainable design
  • Increased productivity due to on-demand cooling

Catalog No. NC3450489


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.