Learn More
Chemglass Life Sciences ECODryBlock for 0.5mL PCR Tubes
SDP
Catalog No. 5000493068
Encompass_Preferred
Designed for small-volume samples, typically employed in molecular biology and biochemistry for DNA, RNA, or protein preparation and analysis.
Catalog No. 50-004-93068 Supplier Chemglass Life Sciences Supplier No. CG1991V109
Designed for small-volume samples, typically employed in molecular biology and biochemistry for DNA, RNA, or protein preparation and analysis.