Learn More
Chemglass Life Sciences ECODryBlock for 1.5mL Microcentrifuge Tubes for 10.8mm to 11mm OD Tubes
SDP
Catalog No. 5000493120
Encompass_Preferred
Designed for small-volume samples, typically employed in molecular biology and biochemistry for DNA, RNA, or protein preparation and analysis.
Catalog No. 50-004-93120 Supplier Chemglass Life Sciences Supplier No. CG1991V110
Designed for small-volume samples, typically employed in molecular biology and biochemistry for DNA, RNA, or protein preparation and analysis.