Learn More
Chemglass Life Sciences ECODryBlock for 12.5mm 2 x 6 Cuvettes, 12.5 Wide x25.4mm Deep, 2 Wells
SDP
Catalog No. 5000493112
Encompass_Preferred
Allow controlled heating of cuvettes for spectrophotometric analysis, supporting kinetic studies, enzyme reactions, or temperature-dependent absorbance measurements.
Catalog No. 50-004-93112 Supplier Chemglass Life Sciences Supplier No. CG1991V131
Allow controlled heating of cuvettes for spectrophotometric analysis, supporting kinetic studies, enzyme reactions, or temperature-dependent absorbance measurements.