Learn More
Chemglass Life Sciences ECODryBlock for 12mm & 13mm Round Bottom Tubes, 13.9mm O.D x 48.4mm Deep Well, 16 Wells
SDP
Catalog No. 5000493087
Encompass_Preferred
Commonly used for general laboratory heating tasks, such as preparing solutions, dissolving substances, or incubating samples in routine chemical and biological analyses.
Catalog No. 50-004-93087 Supplier Chemglass Life Sciences Supplier No. CG1991V102
Commonly used for general laboratory heating tasks, such as preparing solutions, dissolving substances, or incubating samples in routine chemical and biological analyses.