Learn More
Chemglass Life Sciences ECODryBlock for 15mm OD vials like 15mm x 45mm, 4mL (1 Dram) Vials
SDP
Catalog No. 5000493123
Encompass_Preferred
Suitable for reaction vials, often used in chemistry and pharmaceutical applications for compound synthesis, extraction, or sample preparation.
Catalog No. 50-004-93123 Supplier Chemglass Life Sciences Supplier No. CG1991V120
Suitable for reaction vials, often used in chemistry and pharmaceutical applications for compound synthesis, extraction, or sample preparation.