Learn More
Chemglass Life Sciences ECODryBlock for 16mm Round Bottom COD Test Tube, 16mm O.D x 47mm Deep Well, 24 Wells
SDP
Catalog No. 5000493113
Encompass_Preferred
Dedicated to COD (Chemical Oxygen Demand) analysis in environmental testing, ensuring accurate digestion of water and wastewater samples under controlled conditions.
Catalog No. 50-004-93113 Supplier Chemglass Life Sciences Supplier No. CG1991V132
Dedicated to COD (Chemical Oxygen Demand) analysis in environmental testing, ensuring accurate digestion of water and wastewater samples under controlled conditions.